Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ortho-Boric acid
Orthoboric acid
Orthoboric acid
SWEDBO975445
:
SWED HANDLING
:
Orthoboric acid
SWEDBO975445
:
SWED HANDLING
:
Pk:
25 kg
Seq:
4283
:
:
13,01326
:
185 °C (1013 hPa)
:
160 °C
:
1.44 g/cm³ (25 °C)
:
61.83 g/mol
:
Ambient
:
233-139-2
:
01-2119486683-25
:
GHS08
:
10043-35-3
:
B(OH)₃
:
MFCD00011337