4'-Ethoxyacetanilide
Glass bottle for solids, 500 g
Catalog # MOLEM40197688
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD00009094
- Molecular Weight:179.22 g/mol
- EINECS:200-533-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



