3-Aminocrotononitrile cis- and trans-mixture 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1118-61-2
:
Formula:C₄H₆N₂
MDL Number:MFCD00008071
:82.11 g/mol
Storage Temperature:Refrigerator
EINECS:214-266-2
UN:3439
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Yellow Flakes
- Infrared spectrum:Conforms
- GC:≥95 %