Order Entry
Norway
ContactUsLinkComponent
Calcium chloride ≥99%, white powder cell culture reagent
Calcium chloride ≥99%, white powder cell culture reagent
Leverandør:  MP Biomedicals
Sertifikat
Calcium chloride ≥99%, white powder cell culture reagent
Leverandør:  MP Biomedicals

Spesifikasjonstestresultater

  • Identity Test:
    Passes
  • Purity:
    ~99%
  • pH (5% aq soln):
    4 - 9.5
  • Ammonium:
    ≤0.005%
  • Barium:
    ≤0.005%
  • Heavy Metals:
    ≤5ppm
  • Insoluble Matter:
    ≤0.01%
  • Iron:
    ≤5ppm
  • Magnesium:
    ≤0.005%
  • Oxidizing Substances:
    ≤0.003%
  • Potassium:
    ≤0.01%
  • Sodium:
    ≤0.02%
  • Strontium:
    ≤0.1%
  • Sulfate:
    ≤0.01%

Spesifikasjonstestresultater