Sodium hydrogen selenite ≥96.0% (RT), Purum p.a.
Catalog # 71658-100G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:96.0 - 104.0 %
- pH (5 %, 20°C):5.4 - 7.2
- Colour of the solution:complying
- Clearness of the solution:complying
- Calcium (Ca):Max. 100 ppm
- Cadmium (Cd):Max. 50 ppm
- Cobalt (Co):Max. 50 ppm
- Copper (Cu):Max. 100 ppm
- Iron (Fe):Max. 50 ppm
- Potassium (K):Max. 100 ppm
- Nickel (Ni):Max. 50 ppm
- Lead (Pb):Max. 300 ppm
- Zinc (Zn):Max. 100 ppm
- Chloride (Cl):Max. 100 ppm
- Selenate and sulfate (as SO₄):Max. 1500 ppm
Specification Test Results
About this item
:
:150.96 g/mol
Storage Temperature:Ambient
EINECS:231-966-3
UN:2630
ADR:6.1,I
:GHS08
:GHS06
:GHS09
CAS Number:7782-82-3
Formula:HNaO₃Se
MDL Number:MFCD00003525