Capillary columns, BPX50
Supplier: Trajan Scientific and Medical
Certificates
About this item
These columns are suitable for EPA methods 604, 608, 8060, 8081, triazines/herbicides, drug screening, steroids and a variety of pharmaceutical applications.
50% phenyl polysilphenylene-siloxane
Max. temperature: 80 °C to 360/370 °C
Suitable replacement for: OV-17, SP-2250, DB-17, DB-17ms, DB-17ht, Rtx-50, SPB-50, HP-50+, HP-17