GL tube fittings, ground joint, BOLA
PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
- Temperature resistance: −200 to +250 °C
- Universal chemical resistance
- Vacuum: suitable
Fitting made of PTFE, for transition from ground joints to GL threads. For connection of hard-walled tubing (e.g. PTFE, PFA, FEP) with BOLA Laboratory Screw Joints . Ground joint body with turned rings and knurled grip for opening. The product is only exposed to PTFE.