Vann for molekylærbiologi
:7732-18-5
:
:H₂O
:MFCD00011332
:18.02 g/mol
:13,10098
:100 °C (1013 hPa)
:0 °C
:1 g/cm³
:Romtemperatur
:231-791-2
- Appearance:Clear liquid
- Additional info:residue (at bottling): < 0.0001%
- Color scale:≤5 APHA
- Dnase,Rnase,Protease activity:none detected
- Chloride (Cl):≤0.1 ppm
- Sulfate (SO4):≤0.5 ppm
- Lead (Pb):≤1 ppb