Order Entry
Norway
ContactUsLinkComponent
Water, pure, demineralised
Water, pure, demineralised
Certificates
undefined
Water, pure, demineralised
Certificates

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • pH:
    6 to 8
  • Silicon dioxide (SiO2):
    ≤0.1 ppm
  • Conductivity:
    ≤5.0 µS/cm

Specification Test Results

Related Information