

Sodium nitrate 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7631-99-4
:
Formula:NaNO₃
MDL Number:MFCD00011119
:84.99 g/mol
Merck Index:13,08720
Boiling Pt:380 °C (760 mmHg)
Melting Pt:306 °C
Density:2,26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-554-3
UN:1498
ADR:5.1,III
REACH:01-2119488221-41
:GHS07
:GHS03
Specification Test Results
- Appearance:White Crystals
- pH:5.5 to 8.3 (5 % solution at 25.0°C)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤10 ppm
- Iodate (IO3):≤5 ppm
- Nitrite (NO2):≤10 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤30 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤3 ppm
- Magnesium (Mg):≤20 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %