Order Entry
Norway
ContactUsLinkComponent
 
Oxidizing
Lithium nitrate, Sigma-Aldrich®
Catalog # 1.12230.0250
Supplier:  Merck
CAS Number:  
Oxidizing
Lithium nitrate, Sigma-Aldrich®
Catalog # 1.12230.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    522 g/l (20 °C)
  • Melting Point:
    255 °C
  • Molar Mass:
    68.94 g/mol
  • Bulk Density:
    910 kg/m3
  • Density:
    2.36 g/cm3 (20 °C)
  • pH:
    7 - 9 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :68.95 g/mol
Merck Index:13,05558
Melting Pt:255 °C
Density:2.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-218-9
UN:2722
ADR:5.1,
 :M-Clarity™ quality level = MQ200
 :GHS03
CAS Number:7790-69-4
Formula:LiNO₃
MDL Number:MFCD00011094