Stirrer Bearings, glass
Supplier: Bohlender
Certificates
About this item
Combination of borosilicate glass piece with ground joint, an interior PTFE shaft guide with integrated special gasket and a GL screw cap made
of polyphenylene sulphide.
- Suitable for vacuum
- Perfect bearing for stirrer shafts made of stainless steel, glass and for BOLA stirrer shafts
- Temperature resistance: –15 to +200 °C
- Chemical resistance
The special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap provide a good sealing of the stirrer shaft.
This gasket can be exchanged after wearing.
Certifications: FDA conform