Virginal rods, BOLA
Virginal rods made of white PTFE.
- Diameter and length are nominal dimensions and can contain a machining allowance
- Temperature resistance from −200 to +250 °C
- Universal chemical resistance
- FDA conform
Virginal rods made of white PTFE.