Rubidiumklorid analytisk reagens, Supelco®
Catalog # 1.07615.0025
Supplier: Merck
CAS Number:
Specifications
- Pakketype:Glass bottle
- Fp:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:910 g/l (20 °C)
- Melting Point:715 °C
- Molar Mass:120.92 g/mol
- Bulk Density:1000 - 1200 kg/m3
- Boiling Point:1390 °C
- Density:2.76 g/cm3 (20 °C)
- pH:(50 g/l, H2O, 20 °C) neutral
Specification Test Results
About this item
:
:120.92 g/mol
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:715 °C
Density:2,8 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Romtemperatur
EINECS:232-240-9
CAS Number:7791-11-9
Formula:RbCl
MDL Number:MFCD00011187