Rubidium chloride ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-11-9
:
Formula:RbCl
MDL Number:MFCD00011187
:120.92 g/mol
Merck Index:14,8284
Boiling Pt:1390 °C
Melting Pt:718 °C
Density:2.800
Storage Temperature:Ambient
EINECS:232-240-9