Overhead stirrer tools, radial stirrers, six paddles, BOLA
Supplier: Bohlender
Certificates
About this item
Stirrer shafts made of PTFE-jacketed stainless steel and the impeller completely made of PTFE with six radial paddles.
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Axial suction of mixture, strong radial flow. Ideal for aerating liquids.