Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Specification Test Results
- Assay:Min. 99 %
- Hexacyanoferrates (as K4[Fe(CN)6]):Max. 15 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Br (Bromide):Max. 0.02 %
- I (Iodide):Max. 100 ppm
- Fe (Iron):Max. 10 ppm