Order Entry
Norway
ContactUsLinkComponent
Sodium chloride, Suprapur® for trace analysis, Supelco®
Sodium chloride, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Sodium chloride, Suprapur® for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    358 g/l (20 °C)
  • Melting Point:
    801 °C
  • Molar Mass:
    58.44 g/mol
  • Bulk Density:
    1140 kg/m3
  • Boiling Point:
    1461 °C (1013 hPa)
  • Vapor Pressure:
    1.3 hPa (865 °C)
  • Density:
    2.17 g/cm3 (20 °C)
  • pH:
    6 - 9 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information