Order Entry
Norway
ContactUsLinkComponent
 
Sodium chloride ≥99.9%, HiPerSolv CHROMANORM® for HPLC
  153274V
 :  
undefined
Sodium chloride ≥99.9%, HiPerSolv CHROMANORM® for HPLC
  153274V
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay (argentometric, after ignition):
    Min. 99.9 %
  • Identity:
    Passes test
  • pH-value (5 %; water):
    5.0 - 8.0
  • Transmission (206 nm; 1 mol/L; 1 cm):
    Min. 10 %
  • Transmission (254 nm; 1 mol/L; 1 cm):
    Min. 95 %
  • Br (Bromide):
    Max. 0.005 %
  • I (Iodide):
    Max. 0.001 %
  • PO4 (Phosphate):
    Max. 0.0005 %
  • SO4 (Sulphate):
    Max. 0.001 %
  • Fe (Iron):
    Max. 0.0001 %
  • K (Potassium):
    Max. 0.005 %
  • Pb (Lead):
    Max. 0.0002 %

 

 

 :
 :58.44 g/mol
 :14,08599
 :1413 °C (1013 hPa)
 :801 °C
 :2.16 g/cm³ (20 °C)
 :Ambient
 :231-598-3
 :7647-14-5
 :NaCl
 :MFCD00003477