Order Entry
Norway
ContactUsLinkComponent
Sodium chloride ≥98.5% for molecular biology
Sodium chloride ≥98.5% for molecular biology
Supplier:  PanReac AppliChem
Certificates
Sodium chloride ≥98.5% for molecular biology
Supplier:  PanReac AppliChem
Certificates

Specification Test Results

  • DNases/RNases/Proteases:
    not detectable
  • Assay (titr.):
    min. 99.5 %
  • pH (5 %; H2O; 20°C):
    5.0 - 8.0
  • Heavy metals (as Pb):
    max. 0.0005 %
  • Bromide:
    max. 0.005 %
  • Iodide:
    max. 0.001 %
  • Phosphate:
    max. 0.001 %
  • Sulfate:
    max. 0.01 %
  • Ba:
    max. 0.001 %
  • Ca:
    max. 0.002 %
  • Fe:
    max. 0.0001 %
  • K:
    max. 0.01 %
  • MG:
    max. 0.001 %

Specification Test Results