Potassium chloride 99.0-100.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Merck Index:14,7621
Boiling Pt:1500 °C (subl.)
Melting Pt:773 °C
Density:1.984
Storage Temperature:Ambient
EINECS:231-211-8