Order Entry
Norway
ContactUsLinkComponent
Cesium chloride
Cesium chloride
Catalog # SHETIB37060
Supplier:  IBI SCIENTIFIC
CAS Number:  
Cesium chloride
Catalog # SHETIB37060
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:646 °C
Density:3.7808 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-600-2
CAS Number:7647-17-8
Formula:CsCl
MDL Number:MFCD00010955