Order Entry
Norway
ContactUsLinkComponent
 
Cesium chloride, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  2012.0100
 :  Avantor
 :  
Cesium chloride, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  2012.0100
 :  Avantor
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Assay:
    min. 99.5 %
  • Nitrogen Compounds (as N):
    max. 0.001 %
  • Aluminium (Al):
    max. 5
  • Iron (Fe):
    max. 3
  • Potassium (K):
    max. 20
  • Magnesium (Mg):
    max. 5
  • Sodium (Na):
    max. 20
  • Heavy Metals (as Pb):
    max. 5
  • Sulfate (SO4):
    max. 50

 

 

 :
 :168.36 g/mol
 :13,02021
 :1303 °C (1013 hPa)
 :646 °C
 :3.7808 g/cm³ (20 °C)
 :Ambient
 :231-600-2
 :7647-17-8
 :CsCl
 :MFCD00010955