Sodium hydrogen carbonate 99.0-101.0% Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Merck Index:13,08655
Melting Pt:270 °C
Density:2.159 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-633-8
Specification Test Results
- Assay:99.0 - 101.0 %
- Appearance::.
- White or almost white crystalline powder:Passes test
- Identification A:Passes test
- Identification B:Passes test
- Identification (Na):Passes test
- Appearance of solution:Passes test
- CO3 (Carbonate):Passes test
- Cl (Chloride):≤ 150 ppm
- SO4 (Sulphate):≤ 150 ppm
- NH4 (Ammonium):≤ 20 ppm
- Ca (Calcium):≤ 100 ppm
- Fe (Iron):≤ 20 ppm
- Heavy metals (as Pb):≤ 10 ppm
- Residual solvents:Unlikely by manuf.process
- As (Arsenic):≤ 2 ppm
- Cd (Cadmium):≤ 1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 2 ppm
- Ni (Nickel):≤ 1 ppm
- Pb (Lead):≤ 3 ppm
- V (Vanadium):≤ 1 ppm
- Conforms to Ph.Eur.:Passes test
- Original manufacturer code:R or B
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
27775_SODIUM HYDROGEN CARBONATE PH.EUR._BSE_REV0
BSE/TSE Certificates
27775_SODIUM HYDROGEN CARBONATE PH.EUR._LATEX_REV0 Sodium Hydrogen Carbonate 99 - 101% Ph. Eur.
Additional Product Details
spec_VWRC27775.293 spec_VWRC27775.293 spec_VWRC27775.362 spec_VWRC27775.362 spec_VWRC27775.464
Product specification
VWR Chemicals for Biopharma and Pharma Production