kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Sodium hydrogen carbonate
Sodium hydrogen carbonate 99.7-100.3% ACS
Sodium hydrogen carbonate 99.7-100.3% ACS
:
Thermo Fisher Scientific
Sodium hydrogen carbonate 99.7-100.3% ACS
:
Thermo Fisher Scientific
:
144-55-8
:
:
NaHCO₃
:
MFCD00003528
:
84.01 g/mol
:
14,8583
:
270 °C
:
2.159
:
Ambient
:
205-633-8
msds_ALFA014707
Thermo Scientific Acroseal 25 Years