
Potassium carbonate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:584-08-7
:
Formula:K₂CO₃
MDL Number:MFCD00011382
:138.21 g/mol
Merck Index:13,07702
Melting Pt:891 °C
Density:2.42 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1120 g/l (20 °C)
- Melting Point:891 °C
- Molar Mass:138.21 g/mol
- Bulk Density:750 kg/m3
- Density:2.43 g/cm3 (20 °C)
- pH:11.5 - 12.5 (50 g/l, H2O, 20 °C)