



Potassium cyanide 97+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:151-50-8
:
Formula:KCN
MDL Number:MFCD00011397
:65.12 g/mol
Merck Index:13,07709
Boiling Pt:1625 °C
Melting Pt:634 °C
Density:1,55 g/cm³ (20 °C)
EINECS:205-792-3
UN:1680
ADR:6.1,I
:GHS06
:GHS09
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥97.0 %
- Water:≤0.2 %
- Carbonate (CO3):≤1 % (as K2CO3)
- Potassium formate:≤0.5 %
- Free alkali (KOH):≤1 %