
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Catalog # RESERES3007T-A102X
Supplier: RESEARCH ORGANICS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:229.25 g/mol
EINECS:230-906-3
:GHS07
Storage Temperature:Ambient
CAS Number:7365-44-8
Formula:C₆H₁₅NO₆S
MDL Number:MFCD00007532