Order Entry
Norway
ContactUsLinkComponent
 
 
 
Pyridinium tribromide, Sigma-Aldrich®
  8.14345.0050
 :  Merck
 :  
 
 
Pyridinium tribromide, Sigma-Aldrich®
  8.14345.0050
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

 

 

  • Solubility:
    0.17 g/l (20 °C)
  • Melting Point:
    -8 °C
  • Molar Mass:
    183.12 g/mol
  • Boiling Point:
    240 - 242 °C (1013 hPa)
  • Vapor Pressure:
    0.1 hPa (20 °C)
  • Flash Point:
    118 °C
  • Refractive Index:
    1.457 (25 °C, 589 nm)
  • Density:
    1.03 g/cm3 (20 °C)
  • Ignition Point:
    215 °C

 

 

 :
 :319.82 g/mol
 :8,II
 :M-Clarity™ quality level = MQ200
 :254-446-8
 :3261
 :GHS05
 :GHS07
 :Refrigerator
 :39416-48-3
 :C₅H₆Br₃N
 :MFCD00013223