Order Entry
Norway
ContactUsLinkComponent
Hydrochloric acid 36%, Supelco®
Hydrochloric acid 36%, Supelco®
Catalog # 1.15186.1000
Supplier:  Merck
CAS Number:  
Hydrochloric acid 36%, Supelco®
Catalog # 1.15186.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Seq:
    5037

Specifications

About this item

 :
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,
 :M-Clarity™ quality level = MQ100
 :36.46 g/mol
EINECS:231-595-7
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324