Organic reference standard, Furan 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA13965000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:110-00-9
- Description:Furan 100 µg/ml in acetonitrile
- Formula:C₄H₄O
- Description:Furan