Cyclohexanol for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:108-93-0
:
Formula:C₆H₁₂O
MDL Number:MFCD00003855
:100.16 g/mol
Merck Index:13,02754
Boiling Pt:161 °C (1013 hPa)
Melting Pt:25 °C
Density:0.947 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:59 °C
EINECS:203-630-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:40 g/l (20 °C)
- Melting Point:25 °C
- Molar Mass:100.16 g/mol
- Boiling Point:161 °C (1013 hPa)
- Vapor Pressure:0.6 hPa (20 °C)
- Refractive Index:1.4656 (20 °C, 589 nm)
- Explosion Limit:2 - 11.2 %(V)
- Density:0.94 g/cm3 (25 °C)
- pH:6.5 (40 g/l, H2O, 20 °C)
- Ignition Point:300 °C
- Saturation concentration:5 g/m3 (20 °C) Air