
Quinaldine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:91-63-4
:
Formula:C₁₀H₉N
MDL Number:MFCD00006756
:143.19 g/mol
Merck Index:12,08228
Boiling Pt:247.6 °C (1013 hPa)
Melting Pt:–2 °C
Density:1.0585 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:79 °C
EINECS:202-085-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:2.5 g/l (20 °C)
- Melting Point:-2 °C
- Molar Mass:143.18 g/mol
- Boiling Point:246.9 °C
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:79 °C
- Refractive Index:1.6126 (20 °C, 589 nm)
- Density:1.06 g/cm3 (20 °C)
- pH:6.9 (H2O, 20 °C) (saturated aqueous solution)
Specification Test Results
Frequently Bought Together