Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-4-quinolinol
2-Methyl-4-quinolinol
2-Methyl-4-quinolinol
2-Methyl-4-quinolinol
Supplier:
Thermo Scientific
Certificates
Expand image
2-Methyl-4-quinolinol
Supplier:
Thermo Scientific
About this item
CAS Number:
607-67-0
:
Formula:
C₁₀H₉NO
MDL Number:
MFCD00006758
:
159.19 g/mol
Boiling Pt:
313 °C (1013 hPa)
EINECS:
210-140-6
:
GHS07