

Benzyl benzoate
Catalog # EHERC10537700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:212.25 g/mol
Merck Index:13,01129
Boiling Pt:323 °C (1013 hPa)
Melting Pt:17…20 °C
Density:1.118 g/cm³ (20 °C)
Flash Pt:158 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-402-9
UN:3082
ADR:9,III
:GHS07
:GHS09
CAS Number:120-51-4
Formula:C₁₄H₁₂O₂
MDL Number:MFCD00003075