Acenaphthene-[D10] 2000 µg/ml in dichloromethane, Supelco®
Catalog # 48417.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- CAS No.:15067-26-2
- Description:Acenaphthene-[D10] 2,000 µg/ml in dichloromethane
- Formula:C₁₂D₁₀
- Description:Acenaphthene-[D10]