Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Boc-Pro-OH

Plastic bottle for solids, 100 g
Catalog # 04-12-0010-100
Boc-Pro-OH
Catalog # 04-12-0010-100
CAS Number:  15761-39-4
Boc-Pro-OH

Plastic bottle for solids, 100 g

CAS Number:  15761-39-4
Price
kr 1 125,00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
  • CAS Number:15761-39-4
  • Formula:C₁₀H₁₇NO₄
  • MDL Number:MFCD00037324
  • Molecular Weight:215.25 g/mol
  • EINECS:239-848-3
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications