Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methyl-2-thenoic acid
4-Methyl-2-thenoic acid
4-Methyl-2-thenoic acid
4-Methyl-2-thenoic acid
Supplier:
Thermo Scientific
Certificates
Expand image
4-Methyl-2-thenoic acid
Supplier:
Thermo Scientific
About this item
CAS Number:
14282-78-1
:
Formula:
C₆H₆O₂S
MDL Number:
MFCD00089284
:
142.18 g/mol