


Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:4 °C
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:0.52 g/l (20 °C)
- Melting Point:-95 °C
- Molar Mass:92.14 g/mol
- Boiling Point:110.6 °C (1013 hPa)
- Vapor Pressure:29 hPa (20 °C)
- Flash Point:4 °C
- Refractive Index:1.4968 (20 °C)
- Explosion Limit:1.2 - 8 %(V)
- Density:0.87 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:535 °C
- Viscosity kinematic:0.7 mm2/s (20 °C)
- Synonyms:Toluol, Methylbenzene
- Hill Formula:C7H8
- Chemical Formula:C6H5CH3