Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
Norway
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
ContactUsLinkComponent
Home
tert-Butylbenzene
tert-Butylbenzene
tert-Butylbenzene
tert-Butylbenzene
EHERC10931100
:
EHRENSTORFER
:
tert-Butylbenzene
EHERC10931100
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
1 mL
Seq:
2070
:
:
134.22 g/mol
:
13,01550
:
169 °C (1013 hPa)
:
–58 °C
:
0.8662 g/cm³ (20 °C)
:
44 °C
:
Ambient
:
202-632-4
:
2709
:
3,III
:
98-06-6
:
C₁₀H₁₄
:
MFCD00008816