Order Entry
Norway
ContactUsLinkComponent
4-Ethylphenol
Irritant
4-Ethylphenol
Catalog # EHERC13352000
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
4-Ethylphenol
Catalog # EHERC13352000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

About this item

 :
 :122.17 g/mol
Boiling Pt:218 °C (1013 hPa)
Melting Pt:40…43 °C
Density:1.011 g/cm³ (20 °C)
Flash Pt:100 °C
Storage Temperature:Ambient
EINECS:204-598-6
UN:2430
ADR:8,III
 :GHS07
CAS Number:123-07-9
Formula:C₈H₁₀O
MDL Number:MFCD00002393