Order Entry
Norway
ContactUsLinkComponent
Safety gas tubing
Safety gas tubing
Catalog # BOCH7835
Supplier:  BOCHEM
undefined
Safety gas tubing
Catalog # BOCH7835
Supplier:  BOCHEM

Specifications

  • Tubing int.Ø:
    10 mm
  • Wall thickness:
    2 mm
  • Description:
    Safety gas tubing
  • Length:
    1 m
  • Pk:
    1 m

Specifications

About this item