

4-Fluoro-3-nitrobenzotrifluoride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:367-86-2
:
Formula:C₇H₃F₄NO₂
MDL Number:MFCD00007059
:209.1 g/mol
Boiling Pt:91…92 °C (15 mmHg)
Density:1.494
Flash Pt:90 °C (194 °F)
Storage Temperature:Ambient
EINECS:206-702-5
UN:2810
ADR:6.1,III
:GHS06
:GHS07