Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-iodotoluene
2-Chloro-4-iodotoluene
2-Chloro-4-iodotoluene
2-Chloro-4-iodotoluene
Supplier:
Thermo Scientific
Certificates
Utvid bildet
2-Chloro-4-iodotoluene
Supplier:
Thermo Scientific
About this item
CAS Number:
83846-48-4
:
Formula:
C₇H₆ClI
MDL Number:
MFCD00040872
:
252.48 g/mol
EINECS:
281-024-0