Acetophenone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-86-2
:
Formula:C₈H₈O
MDL Number:MFCD00008724
:120.15 g/mol
Merck Index:14,73
Boiling Pt:201…203 °C
Melting Pt:19…20 °C
Density:1.030
Flash Pt:76 °C (169 °F)
Storage Temperature:Ambient
EINECS:202-708-7
UN:3334
ADR:9,
:GHS07