Order Entry
Norway
ContactUsLinkComponent
Tetrachloroethylene, Supelco®
Tetrachloroethylene, Supelco®
Catalog # 15889-10ML-F
Supplier:  Merck
CAS Number:  
undefined
Tetrachloroethylene, Supelco®
Catalog # 15889-10ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :165.83 g/mol
Merck Index:12,09332
Boiling Pt:121.1 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
 :GHS08
 :GHS09
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834