
3,5-Dibromotoluene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1611-92-3
:
Formula:C₇H₆Br₂
MDL Number:MFCD00013528
:249.93 g/mol
Boiling Pt:246 °C
Melting Pt:37…41 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient