Order Entry
Norway
ContactUsLinkComponent
Bunsen burners, with stopcock
Bunsen burners, with stopcock
Catalog # 451-0005
Supplier:  BOCHEM
undefined
Bunsen burners, with stopcock
Catalog # 451-0005
Supplier:  BOCHEM
Specifications
  • Max. temp.:
    1300 °C
  • Description:
    Bunsen multi gas bunsen burner with stopcock and air regulation
  • Pk:
    1
  • For:
    All gases
Specifications
About this item
This multigas burner can reach a maximum temperature of 1300 °C and has an anti-slip rubber coating under the base.
  • With air regulation