Serotonin hydrochloride, pale gray powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:153-98-0
:
Formula:C₁₀H₁₃ClN₂O
MDL Number:MFCD00012686
:212.68 g/mol
Melting Pt:147…153 °C (lit.)
Storage Temperature:Freezer
ADR:6.1,III
:GHS07
Specification Test Results
- Identity Test:Passes
- FTIR Conforms to Standard:Loss on Drying
- Loss on Drying:≤1%