Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
Dicyclopropyl ketone
Dicyclopropyl ketone
Dicyclopropyl ketone
Dicyclopropyl ketone
MAYBDSHS00118.1
:
Thermo Scientific
:
Dicyclopropyl ketone
MAYBDSHS00118.1
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
110.16 g/mol
:
1224
:
3,III
:
1121-37-5
:
C₇H₁₀O
:
MFCD00001276